Learn More
Waters Corp Glycan Sample Preparation Kit, Reductive Amination, Single Use, 10 Reactions
Catalog No. 502392956 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks Glycan Sample Preparation Kits are designed to work synergistically with the ACQUITY UPLC Glycan Application Solution to produce high-quality data from analyses of pre-column derivatized, labeled glycans. This kit utilizes HILIC Solid Phase Extraction (SPE) material to separate released glycans from proteins and labeled glycans from excess free label. This specific kit includes 20 HILIC SPE sorbents in 1cc cartridges for 10 reactions, suitable for single-use or low-sample volume environments.
  • Developed and rigorously tested to work with ACQUITY UPLC Glycan Application Solution.
  • Utilizes HILIC SPE material for efficient separation of released glycans and labeled glycans.
  • Suitable for single use / low sample volume environments.
  • Includes 1 mg vial of RapiGest SF to assist in protein deglycosylation.
  • Includes tested and robust kit protocols.
Catalog No. 50-239-2956 Supplier Waters Corp Supplier No. 176003119
The GlycoWorks Glycan Sample Preparation Kits are designed to work synergistically with the ACQUITY UPLC Glycan Application Solution to produce high-quality data from analyses of pre-column derivatized, labeled glycans. This kit utilizes HILIC Solid Phase Extraction (SPE) material to separate released glycans from proteins and labeled glycans from excess free label. This specific kit includes 20 HILIC SPE sorbents in 1cc cartridges for 10 reactions, suitable for single-use or low-sample volume environments.
  • Developed and rigorously tested to work with ACQUITY UPLC Glycan Application Solution.
  • Utilizes HILIC SPE material for efficient separation of released glycans and labeled glycans.
  • Suitable for single use / low sample volume environments.
  • Includes 1 mg vial of RapiGest SF to assist in protein deglycosylation.
  • Includes tested and robust kit protocols.