Learn More
Waters Corp Waters GlycoWorks RFMS Clean-up Module, 96 Samples, HILIC µElution Plate, Low Elution Volume
Catalog No. 501462017 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GlycoWorks RapiFluor-MS Clean-up Module supports RapiFluor-MS customers by providing materials for the clean-up of 96 samples. It removes contaminants like salts and detergents from hydrophilic analytes prior to mass spectrometry, allowing for elution volumes as low as 25 µL and eliminating evaporation steps.
  • Supports separate ordering of the clean-up module.
  • Contains materials for 96 samples.
  • Includes Elution Buffer, Sample Diluent, and a HILIC µElution Plate.
  • Effectively removes contaminants from hydrophilic analytes.
  • Enables low elution volumes as low as 25 µL, avoiding evaporation steps.
  • Enhances sensitivity and accuracy of glycan analysis.
Catalog No. 50-146-2017 Supplier Waters Corp Supplier No. 186008913
The GlycoWorks RapiFluor-MS Clean-up Module supports RapiFluor-MS customers by providing materials for the clean-up of 96 samples. It removes contaminants like salts and detergents from hydrophilic analytes prior to mass spectrometry, allowing for elution volumes as low as 25 µL and eliminating evaporation steps.
  • Supports separate ordering of the clean-up module.
  • Contains materials for 96 samples.
  • Includes Elution Buffer, Sample Diluent, and a HILIC µElution Plate.
  • Effectively removes contaminants from hydrophilic analytes.
  • Enables low elution volumes as low as 25 µL, avoiding evaporation steps.
  • Enhances sensitivity and accuracy of glycan analysis.