Learn More
Waters Corp GlycoWorks RFMS N-Glycan Starter Kit, 96 analyses, Sample Preparation Workflow
Catalog No. 502391864 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Re-imagine a streamlined, fast, and highly sensitive way to analyze released N-glycans. The GlycoWorks RapiFluor-MS N-Glycan Kit simplifies sample preparation into just 3 easy steps in as little as 30 minutes, eliminating complicated and time-consuming processes. The unique chemical attributes of RapiFluor-MS (RFMS) provide increased fluorescence quantification and superior mass spectral response. This allows for the use of a single label to gather valuable information, from characterization to routine monitoring, making glycan analysis easier and more sensitive.
  • GlycoWorks deglycosylation module
  • GlycoWorks labeling module
  • GlycoWorks clean-up module
  • GlycoWorks sample collection module
  • ACQUITY UPLC glycan BEH amide column, 130Å, 1.7μm, 2.1 x 150 mm
  • 5050 mM ammonium formate solution—glycan analysis
Catalog No. 50-239-1864 Supplier Waters Corp Supplier No. 176004083
Add to Cart
Add to Cart
Request a Quote
Re-imagine a streamlined, fast, and highly sensitive way to analyze released N-glycans. The GlycoWorks RapiFluor-MS N-Glycan Kit simplifies sample preparation into just 3 easy steps in as little as 30 minutes, eliminating complicated and time-consuming processes. The unique chemical attributes of RapiFluor-MS (RFMS) provide increased fluorescence quantification and superior mass spectral response. This allows for the use of a single label to gather valuable information, from characterization to routine monitoring, making glycan analysis easier and more sensitive.
  • GlycoWorks deglycosylation module
  • GlycoWorks labeling module
  • GlycoWorks clean-up module
  • GlycoWorks sample collection module
  • ACQUITY UPLC glycan BEH amide column, 130Å, 1.7μm, 2.1 x 150 mm
  • 5050 mM ammonium formate solution—glycan analysis