Learn More
Waters Corp GlycoWorks Sample Collection Module, 1 mL, 600 μL, Automation
Catalog No. 502391913 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GlycoWorks Sample Collection Module provides the necessary components for efficient sample cleanup, minimizing sample transfer during glycan analysis workflows. It includes specialized tubes, tube inserts, and cap strips designed for deglycosylation, labeling reactions, and solid phase extraction (SPE) eluate collection. The module is suitable for automated processes and ensures proper heating when used with compatible heat blocks.
  • Includes 15 1 mL tubes for deglycosylation and labeling reactions.
  • Includes 15 600 μL tube inserts for SPE eluate collection.
  • Contains 15 cap strips with slits for direct injection.
  • Includes one 1 mL round collection tray.
  • Includes one waste tray.
  • Tubes are customized to ensure proper heating with compatible heat blocks.
  • Can be stored at room temperature.
  • Unopened products have a minimum shelf life of 1 year.
Catalog No. 50-239-1913 Supplier Waters Corp Supplier No. 186008823
This GlycoWorks Sample Collection Module provides the necessary components for efficient sample cleanup, minimizing sample transfer during glycan analysis workflows. It includes specialized tubes, tube inserts, and cap strips designed for deglycosylation, labeling reactions, and solid phase extraction (SPE) eluate collection. The module is suitable for automated processes and ensures proper heating when used with compatible heat blocks.
  • Includes 15 1 mL tubes for deglycosylation and labeling reactions.
  • Includes 15 600 μL tube inserts for SPE eluate collection.
  • Contains 15 cap strips with slits for direct injection.
  • Includes one 1 mL round collection tray.
  • Includes one waste tray.
  • Tubes are customized to ensure proper heating with compatible heat blocks.
  • Can be stored at room temperature.
  • Unopened products have a minimum shelf life of 1 year.