Learn More
Aviva Systems Biology Corp GLYCTK Antibody - N-terminal region (ARP47720_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47720_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GLYCTK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLYCTK. Alternate names: GLYCTK1, HBEBP2, HBeAgBP4A, HBEBP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59652