Learn More
Aviva Systems Biology Corp GLYCTK Antibody - middle region (ARP47721_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47721_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLYCTK. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GLYCTK. Alternate names: GLYCTK1, HBEBP2, HBeAgBP4A, HBEBP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-285-323