Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-D-leucine 99.0+%, TCI America™
SDP

Catalog No. G0130100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0130100MG 100 mg
G01301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0130100MG Supplier TCI America Supplier No. G0130100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 688-13-1
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00066050
InChI Key DKEXFJVMVGETOO-UHFFFAOYNA-N
Synonym H-Gly-D-Leu-OH
PubChem CID 1715077
IUPAC Name 2-(2-aminoacetamido)-4-methylpentanoic acid
SMILES CC(C)CC(NC(=O)CN)C(O)=O

Specifications

Melting Point 234°C
Color White
Quantity 100 mg
Formula Weight 188.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-D-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.