Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-DL-alanine 99.0+%, TCI America™
SDP

Catalog No. G01121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01121G Supplier TCI America Supplier No. G01121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 926-77-2
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00025587
InChI Key VPZXBVLAVMBEQI-UHFFFAOYSA-N
Synonym H-Gly-DL-Ala-OH
PubChem CID 98153
IUPAC Name 2-[(2-aminoacetyl)amino]propanoic acid
SMILES CC(C(=O)O)NC(=O)CN

Specifications

Melting Point 235°C
Color White
Quantity 1 g
Formula Weight 146.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-DL-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.