Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-DL-leucine 98.0+%, TCI America™
SDP

Catalog No. G01311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01311G Supplier TCI America Supplier No. G01311G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity dipeptide reagent with greater than 98.0% purity for biochemical research.

Chemical Identifiers

CAS 688-14-2
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.227
MDL Number MFCD00066051
InChI Key DKEXFJVMVGETOO-UHFFFAOYSA-N
Synonym H-Gly-DL-Leu-OH
PubChem CID 102468
IUPAC Name 2-[(2-aminoacetyl)amino]-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)NC(=O)CN

Specifications

Color White
Packaging Glass Bottle
Quantity 1 g
Formula Weight 188.23
Percent Purity ≥98.0% (T)
Grade Biochemical
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-DL-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.