Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-DL-norvaline 97.0+%, TCI America™
SDP

Catalog No. G01341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01341G Supplier TCI America Supplier No. G01341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dipeptide reagent with 97.0% minimum purity determined by HPLC and nonaqueous titration.

Chemical Identifiers

CAS 2189-27-7
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.2
MDL Number MFCD00038186
InChI Key JXIQKLAZYWZTRA-UHFFFAOYSA-N
Synonym H-Gly-DL-Nva-OH
PubChem CID 86841
IUPAC Name 2-[(2-aminoacetyl)amino]pentanoic acid
SMILES CCCC(C(=O)O)NC(=O)CN

Specifications

Color White
Quantity 1 g
Formula Weight 174.20
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-DL-norvaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.