Learn More
Medchemexpress LLC Glycyl-DL-phenylalanine | 721-66-4 | MFCD00065109 | 97.0% | 222.24 g/mol | C11H14N2O3 | 5 G
SDP
Catalog No. 5000303996
Encompass_Preferred
Glycyl-DL-phenylalanine is an amino acid dipeptide derivative used in biochemical research as a reference compound and building block. It is supplied as a solid and is commonly used in enzymatic assays, peptide synthesis studies, and analytical method development.
  • Solid form for convenient storage and handling.
  • Verified purity by NMR (assay 97.0%).
  • Suitable as a reference standard in biochemical and pharmacological studies.
  • Compatible with peptide synthesis and analytical workflows.
  • Provides a glycyl-phenylalanine motif for enzymatic and binding studies.
Catalog No. 50-003-03996 Supplier Medchemexpress LLC Supplier No. HYW0140005G
Glycyl-DL-phenylalanine is an amino acid dipeptide derivative used in biochemical research as a reference compound and building block. It is supplied as a solid and is commonly used in enzymatic assays, peptide synthesis studies, and analytical method development.
  • Solid form for convenient storage and handling.
  • Verified purity by NMR (assay 97.0%).
  • Suitable as a reference standard in biochemical and pharmacological studies.
  • Compatible with peptide synthesis and analytical workflows.
  • Provides a glycyl-phenylalanine motif for enzymatic and binding studies.