Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-DL-phenylalanine, TCI America™
SDP

Catalog No. G01351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01351G Supplier TCI America Supplier No. G01351G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Peptide reagent with purity as determined by nonaqueous titration.

Chemical Identifiers

CAS 721-66-4
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.24
MDL Number MFCD00065110,MFCD00065109
InChI Key JBCLFWXMTIKCCB-UHFFFAOYNA-N
Synonym H-Gly-DL-Phe-OH
PubChem CID 97415
IUPAC Name 2-(2-aminoacetamido)-3-phenylpropanoic acid
SMILES NCC(=O)NC(CC1=CC=CC=C1)C(O)=O

Specifications

Color White
Packaging Bottle
Quantity 1 g
Formula Weight 222.24
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-DL-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.