Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-DL-tryptophan 98.0+%, TCI America™
SDP

Catalog No. G0190100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0190100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G0190100MG Supplier TCI America Supplier No. G0190100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dipeptide reagent suitable for biochemical research applications.

Chemical Identifiers

CAS 2189-26-6
Molecular Formula C13H15N3O3
Molecular Weight (g/mol) 261.281
MDL Number MFCD00022767
InChI Key AJHCSUXXECOXOY-UHFFFAOYSA-N
Synonym H-Gly-DL-Trp-OH
PubChem CID 102763
IUPAC Name 2-[(2-aminoacetyl)amino]-3-(1H-indol-3-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)NC(=O)CN

Specifications

Color Red-White
Quantity 100 mg
Formula Weight 261.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-DL-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.