Learn More
Medchemexpress LLC Glycyl-Exatecan-d5 hydrochloride | 1883333-57-0 | 99.0% | 1 MG
SDP
Catalog No. 5000338395
Encompass_Preferred
Glycyl-Exatecan-d5 hydrochloride is a deuterium-labeled derivative of Glycyl-Exatecan, which is itself a derivative of Exatecan. This compound functions as an anticancer agent, demonstrating significant antitumor activity. It is suitable for research related to various cancers, including solid tumors. It also serves as a tracer and an internal standard for quantitative analytical methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Derivative of Exatecan
  • Anticancer agent with significant antitumor activity
  • Suitable for solid tumors research
  • Functions as a tracer
  • Used as an internal standard for quantitative analysis
Catalog No. 50-003-38395 Supplier Medchemexpress LLC Supplier No. HY128995AS1MG
Glycyl-Exatecan-d5 hydrochloride is a deuterium-labeled derivative of Glycyl-Exatecan, which is itself a derivative of Exatecan. This compound functions as an anticancer agent, demonstrating significant antitumor activity. It is suitable for research related to various cancers, including solid tumors. It also serves as a tracer and an internal standard for quantitative analytical methods such as NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Derivative of Exatecan
  • Anticancer agent with significant antitumor activity
  • Suitable for solid tumors research
  • Functions as a tracer
  • Used as an internal standard for quantitative analysis