Learn More
Medchemexpress LLC Glycyl-Exatecan TFA | 1883333-57-0 | 96.9% | 1 MG
SDP
Catalog No. 5000339335
Encompass_Preferred
Glycyl-Exatecan TFA is an anticancer agent derived from Exatecan. It exhibits significant antitumor activity and is suitable for research on cancers such as solid tumors.
  • Anticancer agent
  • Derivative of Exatecan
  • Exhibits significant antitumor activity
  • Utilized in research for various cancers, including solid tumors
  • Store at -20°C, protected from light and under nitrogen
Catalog No. 50-003-39335 Supplier Medchemexpress LLC Supplier No. HY128995A1MG
Glycyl-Exatecan TFA is an anticancer agent derived from Exatecan. It exhibits significant antitumor activity and is suitable for research on cancers such as solid tumors.
  • Anticancer agent
  • Derivative of Exatecan
  • Exhibits significant antitumor activity
  • Utilized in research for various cancers, including solid tumors
  • Store at -20°C, protected from light and under nitrogen