Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycyl-exatecan TFA | 1883333-57-0 | 96.9% | 606.52 g/mol | C28H26F4N4O7 | 5 MG
SDP

Catalog No. 5000262897
Encompass_Preferred

Glycyl-exatecan TFA is a synthetic trifluoroacetate salt derivative of exatecan used as an experimental anticancer compound in research. It is supplied in solid and DMSO solution formats for in vitro and in vivo pharmacology studies; consult supplier documentation and the SDS for handling and storage guidelines.

  • Trifluoroacetate salt form.
  • Molecular formula C28H26F4N4O7.
  • Molecular weight 606.52 g/mol.
  • Purity typically around 96.9%.
  • Available in small-scale sizes and 10 mM DMSO solutions.
  • Intended for research use only in oncology and pharmacology.
  • Handle according to SDS and laboratory safety procedures.

Catalog No. 50-002-62897 Supplier Medchemexpress LLC Supplier No. HY128995A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycyl-exatecan TFA is a synthetic trifluoroacetate salt derivative of exatecan used as an experimental anticancer compound in research. It is supplied in solid and DMSO solution formats for in vitro and in vivo pharmacology studies; consult supplier documentation and the SDS for handling and storage guidelines.

  • Trifluoroacetate salt form.
  • Molecular formula C28H26F4N4O7.
  • Molecular weight 606.52 g/mol.
  • Purity typically around 96.9%.
  • Available in small-scale sizes and 10 mM DMSO solutions.
  • Intended for research use only in oncology and pharmacology.
  • Handle according to SDS and laboratory safety procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.