Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-L-glutamic Acid 98.0+%, TCI America™
SDP

Catalog No. G0123100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0123100MG 100 mg
G01231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0123100MG Supplier TCI America Supplier No. G0123100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7412-78-4
Molecular Formula C7H12N2O5
Molecular Weight (g/mol) 204.182
MDL Number MFCD00008128
InChI Key IEFJWDNGDZAYNZ-BYPYZUCNSA-N
Synonym glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh
PubChem CID 99278
ChEBI CHEBI:73801
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]pentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)NC(=O)CN

Specifications

Melting Point 158°C
Color White
Quantity 100 mg
Formula Weight 204.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycyl-L-glutamic Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.