Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycyl-L-glutamine | 13115-71-4 | MFCD00077124 | 98.0% | 203.20 g·mol⁻¹ | C7H13N3O4 | 500 MG
SDP

Catalog No. 5000226758
Encompass_Preferred

Glycyl-L-glutamine is a research-grade dipeptide composed of glycine and L-glutamine, provided for biochemical and pharmacological studies. It is used as a reference compound or reagent in peptide metabolism and neuropeptide research.

  • High purity: 98.0%.
  • Molecular weight: 203.20 g·mol⁻¹.
  • Chemical formula: C7H13N3O4.
  • CAS number: 13115-71-4.
  • Packaged as 500 mg in a single vial for laboratory use.
  • Water-soluble and suitable for biochemical assays.

Catalog No. 50-002-26758 Supplier Medchemexpress LLC Supplier No. HY117541500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycyl-L-glutamine is a research-grade dipeptide composed of glycine and L-glutamine, provided for biochemical and pharmacological studies. It is used as a reference compound or reagent in peptide metabolism and neuropeptide research.

  • High purity: 98.0%.
  • Molecular weight: 203.20 g·mol⁻¹.
  • Chemical formula: C7H13N3O4.
  • CAS number: 13115-71-4.
  • Packaged as 500 mg in a single vial for laboratory use.
  • Water-soluble and suitable for biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.