Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyl-L-glutamine Monohydrate 98.0+%, TCI America™
SDP

Catalog No. G02511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G02511G Supplier TCI America Supplier No. G02511G
Add to Cart
Add to Cart

Specifications

Purity ≥98.0% (T)
CAS 172669-64-6
Color White
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 221.213
Product Type Glycyl-L-glutamine Monohydrate
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.