Learn More
Cayman Chemical Glycyl-L-Glutamine (hydrate), Purity: ≥98%, CAS: 172669-64-6, 500 mg
SDP
Catalog No. 502911474
Encompass_Preferred
A dipeptide fragment of β-endorphin; decreases reperfusion damage and increases the time to ischemic contracture in isolated rat hearts in a model of cardiac ischemia-reperfusion injury using eight- and 36-week old rats at 5 mM; inhibits morphine-induced respiratory depression at 1-100 nmol; inhibits acquisition of place preference, delays the development of tolerance, and reduces withdrawal intensity in morphine-conditioned rats
Catalog No. 50-291-1474 Supplier Cayman Chemical Supplier No. 35058500 MG
A dipeptide fragment of β-endorphin; decreases reperfusion damage and increases the time to ischemic contracture in isolated rat hearts in a model of cardiac ischemia-reperfusion injury using eight- and 36-week old rats at 5 mM; inhibits morphine-induced respiratory depression at 1-100 nmol; inhibits acquisition of place preference, delays the development of tolerance, and reduces withdrawal intensity in morphine-conditioned rats