Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycyl-L-histidyl-L-lysine | 49557-75-7 | 99.9% | 340.38 | 25 G
SDP

Catalog No. 5000425910
Encompass_Preferred

Glycyl-L-histidyl-L-lysine is a tripeptide consisting of glycine, L-histidine, and L-lysine residues joined in sequence. It is described as a hepatotropic immunosuppressor and shows an anxiolytic effect. Glycyl-L-histidyl-L-lysine and its copper complexes show good skin tolerance. This product is a solid, white to off-white powder, and should be stored at 4°C, protected from light. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light.

  • Purity of 99.89%
  • Molecular formula C14H24N6O4
  • Soluble in H2O at 100 mg/mL (ultrasonic needed)
  • Decreases TNF-α-dependent IL-6 secretion in fibroblasts
  • Stimulates mitotic activity of hepatocytes
  • Suppresses immune reactivity
  • Produces an anxiolytic effect

Catalog No. 50-004-25910 Supplier Medchemexpress LLC Supplier No. 25GHYP0046
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycyl-L-histidyl-L-lysine is a tripeptide consisting of glycine, L-histidine, and L-lysine residues joined in sequence. It is described as a hepatotropic immunosuppressor and shows an anxiolytic effect. Glycyl-L-histidyl-L-lysine and its copper complexes show good skin tolerance. This product is a solid, white to off-white powder, and should be stored at 4°C, protected from light. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also protected from light.

  • Purity of 99.89%
  • Molecular formula C14H24N6O4
  • Soluble in H2O at 100 mg/mL (ultrasonic needed)
  • Decreases TNF-α-dependent IL-6 secretion in fibroblasts
  • Stimulates mitotic activity of hepatocytes
  • Suppresses immune reactivity
  • Produces an anxiolytic effect

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.