Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycyl-L-proline | 704-15-4 | MFCD00020840 | 99.9% | 172.18 g/mol | C7H12N2O3 | 1 ML
SDP

Catalog No. 5000299877
Encompass_Preferred

Glycyl-L-proline (H-Gly-Pro-OH) is a dipeptide endogenous metabolite used as a research reagent. It is investigated in cancer and metabolic-disease studies and has been reported to induce apoptosis in MCF-7 cells and to affect proline transport and collagen/DNA biosynthesis.

  • High purity: 99.9%.
  • Molecular formula: C7H12N2O3.
  • Molecular weight: 172.18 g/mol.
  • CAS number: 704-15-4.
  • Supplied as a solid suitable for preparing 10 mM stock solutions.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-99877 Supplier Medchemexpress LLC Supplier No. HYW01688710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycyl-L-proline (H-Gly-Pro-OH) is a dipeptide endogenous metabolite used as a research reagent. It is investigated in cancer and metabolic-disease studies and has been reported to induce apoptosis in MCF-7 cells and to affect proline transport and collagen/DNA biosynthesis.

  • High purity: 99.9%.
  • Molecular formula: C7H12N2O3.
  • Molecular weight: 172.18 g/mol.
  • CAS number: 704-15-4.
  • Supplied as a solid suitable for preparing 10 mM stock solutions.
  • Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.