Learn More
Medchemexpress LLC Glycyl-L-tryptophan | 2390-74-1 | MFCD00065978 | >98.0% | 261.28 g/mol | C13H15N3O3 | 50 MG
SDP
Catalog No. 5000283716
Encompass_Preferred
H-Gly-Trp-OH (Glycyl-L-tryptophan) is a dipeptide research reagent composed of glycine and L-tryptophan. Supplied as a solid, it is used in peptide synthesis, biochemical assays, and analytical studies. The compound is intended for research use only and is typically supplied with high purity suitable for laboratory applications.
  • Dipeptide composed of glycine and L-tryptophan.
  • Available as a white to off-white solid suitable for laboratory handling.
  • High purity suitable for peptide synthesis and analytical assays.
  • Soluble in common organic solvents and in dilute aqueous solutions; verify solubility for specific protocols.
  • Intended for research use only, not for human or clinical applications.
Catalog No. 50-002-83716 Supplier Medchemexpress LLC Supplier No. HYW14324150MG
H-Gly-Trp-OH (Glycyl-L-tryptophan) is a dipeptide research reagent composed of glycine and L-tryptophan. Supplied as a solid, it is used in peptide synthesis, biochemical assays, and analytical studies. The compound is intended for research use only and is typically supplied with high purity suitable for laboratory applications.
  • Dipeptide composed of glycine and L-tryptophan.
  • Available as a white to off-white solid suitable for laboratory handling.
  • High purity suitable for peptide synthesis and analytical assays.
  • Soluble in common organic solvents and in dilute aqueous solutions; verify solubility for specific protocols.
  • Intended for research use only, not for human or clinical applications.