Learn More
Medchemexpress LLC Glycylglycine | 556-50-3 | 99.9% | 1 KG
SDP
Catalog No. 5000409969
Encompass_Preferred
Glycylglycine is a non-selective glycylglycine dipeptidase substrate and iNOS inhibitor. It can cross the cell membrane by passive diffusion and is hydrolyzed to glycine in the cell, participating in energy metabolism and antioxidant processes.
  • Promotes spermatogonial stem cells (SSCs) proliferation.
  • Inhibits astrocyte overactivation.
  • Reduces nitric oxide (NO) release.
  • Upregulates expression of neurotrophic factors to support nerve myelin repair.
  • Useful for studying male reproductive biology.
  • Useful for studying neurodegenerative diseases.
Catalog No. 50-004-09969 Supplier Medchemexpress LLC Supplier No. HYD08891000G
Glycylglycine is a non-selective glycylglycine dipeptidase substrate and iNOS inhibitor. It can cross the cell membrane by passive diffusion and is hydrolyzed to glycine in the cell, participating in energy metabolism and antioxidant processes.
  • Promotes spermatogonial stem cells (SSCs) proliferation.
  • Inhibits astrocyte overactivation.
  • Reduces nitric oxide (NO) release.
  • Upregulates expression of neurotrophic factors to support nerve myelin repair.
  • Useful for studying male reproductive biology.
  • Useful for studying neurodegenerative diseases.