Learn More
Medchemexpress LLC Glycylglycine | 556-50-3 | 99.9% | 100 G
SDP
Catalog No. 5000410176
Encompass_Preferred
Glycylglycine is a non-selective glycylglycine dipeptidase substrate and iNOS inhibitor. It can cross the cell membrane by passive diffusion and is hydrolyzed to glycine in the cell, where it participates in energy metabolism and antioxidant processes. It is used to study male reproductive biology and neurodegenerative diseases.
  • Promotes spermatogonial stem cells proliferation
  • Inhibits astrocyte overactivation
  • Reduces nitric oxide release
  • Upregulates expression of neurotrophic factors to support nerve myelin repair
Catalog No. 50-004-10176 Supplier Medchemexpress LLC Supplier No. HYD0889100G
Glycylglycine is a non-selective glycylglycine dipeptidase substrate and iNOS inhibitor. It can cross the cell membrane by passive diffusion and is hydrolyzed to glycine in the cell, where it participates in energy metabolism and antioxidant processes. It is used to study male reproductive biology and neurodegenerative diseases.
  • Promotes spermatogonial stem cells proliferation
  • Inhibits astrocyte overactivation
  • Reduces nitric oxide release
  • Upregulates expression of neurotrophic factors to support nerve myelin repair