Learn More
Medchemexpress LLC Glycylglycine | 556-50-3 | 99.9% | 500 G
SDP
Catalog No. 5000410692
Encompass_Preferred
Glycylglycine is a non-selective glycylglycine dipeptidase substrate and iNOS inhibitor. It can cross cell membranes via passive diffusion and is hydrolyzed to glycine within cells, where it contributes to energy metabolism and antioxidant processes. This compound is used in research to promote spermatogonial stem cell proliferation and inhibit astrocyte overactivation.
  • Reduces nitric oxide release
  • Upregulates expression of neurotrophic factors (PDGFA, FGF2, CNTF)
  • Supports nerve myelin repair
  • Used to study male reproductive biology
  • Used to study neurodegenerative diseases (e.g., neuroprotective mechanisms in multiple sclerosis)
Catalog No. 50-004-10692 Supplier Medchemexpress LLC Supplier No. HYD0889500G
Glycylglycine is a non-selective glycylglycine dipeptidase substrate and iNOS inhibitor. It can cross cell membranes via passive diffusion and is hydrolyzed to glycine within cells, where it contributes to energy metabolism and antioxidant processes. This compound is used in research to promote spermatogonial stem cell proliferation and inhibit astrocyte overactivation.
  • Reduces nitric oxide release
  • Upregulates expression of neurotrophic factors (PDGFA, FGF2, CNTF)
  • Supports nerve myelin repair
  • Used to study male reproductive biology
  • Used to study neurodegenerative diseases (e.g., neuroprotective mechanisms in multiple sclerosis)