Learn More
Medchemexpress LLC HY-D0889 5g Medchemexpress, Glycylglycine CAS:556-50-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD08895G
Encompass_Preferred
Medchemexpress, HY-D0889 5g Glycylglycine CAS:556-50-3 Glycylglycine is the simplest of all peptides and could function as a gamma-glutamyl acceptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3206