Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycylglycine, free base, MP Biomedicals™

Catalog No. ICN10185610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10185610 10 g
ICN10185680 100 g
ICN10185691 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10185610 Supplier MP Biomedicals, Inc Supplier No. 0210185610
Add to Cart
Add to Cart

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N

Specifications

Quantity 10 g
Formula Weight 132.1
Physical Form Crystalline
Chemical Name or Material Glycylglycine Free Base
EINECSNumber 209-127-8
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.