Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycylglycine Hydrochloride Monohydrate 98.0+%, TCI America™
SDP

Catalog No. G01251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G01251G Supplier TCI America Supplier No. G01251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13059-60-4
Molecular Formula C4H9ClN2O3
Molecular Weight (g/mol) 168.577
MDL Number MFCD00035438
InChI Key YHBAZQDEMYQPJL-UHFFFAOYSA-N
Synonym H-Gly-Gly-OH.HCl, Diglycine Hydrochloride
PubChem CID 83095
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid;hydrochloride
SMILES C(C(=O)NCC(=O)O)N.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 168.58
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Glycylglycine Hydrochloride Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.