Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycylglycylglycylglycine 97.0+%, TCI America™
SDP

Catalog No. G01271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01271G 1g
G0127100MG 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01271G Supplier TCI America Supplier No. G01271G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Purity ≥97.0% (HPLC,T)
CAS 637-84-3
Color White
Format Crystal-Powder at 20°C
Molecular Weight (g/mol) 246.223
Product Type Glycylglycylglycylglycine
Quantity 1g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.