Learn More
Cayman Chemical Glycyl-L-Proline (trifluoroacetate salt), Purity: ≥95%, CAS: 76397-13-2, 250 mg
SDP
Catalog No. 502911953
Encompass_Preferred
A dipeptide composed of glycine and proline; used in the study of dipeptide transport in enterocyte basolateral membrane and brush-membrane border vesicles
Catalog No. 50-291-1953 Supplier Cayman Chemical Supplier No. 36182250 MG
A dipeptide composed of glycine and proline; used in the study of dipeptide transport in enterocyte basolateral membrane and brush-membrane border vesicles