Learn More
Cayman Chemical Glabridin, Purity: ≥98%, CAS: 59870-68-7, 50 mg
SDP
Catalog No. 502952128
Encompass_Preferred
A prenylated isoflavonoid that is reported to possess antiviral, antimicrobial, anti-inflammatory, antidiabetic, antiatherogenic, antioxidant, antitumor, and estrogen-like properties
Catalog No. 50-295-2128 Supplier Cayman Chemical Supplier No. 1184350 MG
A prenylated isoflavonoid that is reported to possess antiviral, antimicrobial, anti-inflammatory, antidiabetic, antiatherogenic, antioxidant, antitumor, and estrogen-like properties