Learn More
Cayman Chemical Liquiritin, Purity: ≥98%, CAS: 551-15-5, 10 mg
SDP
Catalog No. 502989161
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits corticosterone-induced apoptosis in PC12 cells at 1 and 2 mg/ml; reduces spinal cord levels of IL-1β, IL-6, and TNF-α and alleviates mechanical and cold allodynia, as well as thermal hyperalgesia, in a mouse model of neuropathic pain induced by CCI of the sciatic nerve at 120 mg/kg; decreases the viability of SiHa, CaSki, and HeLa cervical cancer cells and inhibits migration and invasion of SiHa cells in vitro at 40, 60, and 80 μM; reduces tumor growth in a SiHa mouse xenograft model at 10, 20, and 30 mg/kg
Catalog No. 50-298-9161 Supplier Cayman Chemical Supplier No. 3026310 MG
A flavonoid with diverse biological activities; inhibits corticosterone-induced apoptosis in PC12 cells at 1 and 2 mg/ml; reduces spinal cord levels of IL-1β, IL-6, and TNF-α and alleviates mechanical and cold allodynia, as well as thermal hyperalgesia, in a mouse model of neuropathic pain induced by CCI of the sciatic nerve at 120 mg/kg; decreases the viability of SiHa, CaSki, and HeLa cervical cancer cells and inhibits migration and invasion of SiHa cells in vitro at 40, 60, and 80 μM; reduces tumor growth in a SiHa mouse xenograft model at 10, 20, and 30 mg/kg