Learn More
Apexbio Technology LLC Glycyrrhetinic acid 1449-05-4;471-53-4‏ 20mg
SDP
Catalog No. 5000096308
Encompass_Preferred
Glycyrrhetinic acid (CAS 1449-05-4) is a small-molecule inhibitor targeting 11 -hydroxysteroid dehydrogenase It is designed to modulate enzymatic activity thereby influencing corticosteroid metabolism and cortisol bioavailability Glycyrrhetinic acid exerts its biological activity primarily through inhibition of 11 -hydroxysteroid dehydrogenase Based on these pharmacological properties glycyrrhetinic acid holds research potential in anti-inflammatory antiviral and hepatoprotective applications
Catalog No. 50-000-96308 Supplier Apexbio Technology LLC Supplier No. N279520
Glycyrrhetinic acid (CAS 1449-05-4) is a small-molecule inhibitor targeting 11 -hydroxysteroid dehydrogenase It is designed to modulate enzymatic activity thereby influencing corticosteroid metabolism and cortisol bioavailability Glycyrrhetinic acid exerts its biological activity primarily through inhibition of 11 -hydroxysteroid dehydrogenase Based on these pharmacological properties glycyrrhetinic acid holds research potential in anti-inflammatory antiviral and hepatoprotective applications