Learn More
Selleck Chemical LLC Glycyrrhetinic acid
SDP
Catalog No. 5000004683
Encompass_Preferred
Glycyrrhetinic acid is a triterpenoid aglycone component of the natural product glycyrrhizinic acid and possesses remarkable anti-proliferative and apoptosis-inducing activity in various cancer cell lines
Catalog No. 50-000-04683 Supplier Selleck Chemical LLC Supplier No. S64561G
Glycyrrhetinic acid is a triterpenoid aglycone component of the natural product glycyrrhizinic acid and possesses remarkable anti-proliferative and apoptosis-inducing activity in various cancer cell lines