Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glycyrrhisoflavone | 116709-70-7 | 5 MG
SDP

Catalog No. 5000418716
Encompass_Preferred

Glycyrrhisoflavone, an active prenylflavonoid, inhibits α-glucosidase. It is intended for research use only.

  • Active prenylflavonoid
  • Inhibits α-glucosidase
  • For research use only
  • Purity of 99.43%
  • Appearance is off-white to light yellow solid
  • Classified as a flavonoid, isoflavone, phenol, polyphenol
  • Derived from Glycyrrhiza uralensis Fisch.
  • Shows cytotoxicity against human PC3 cells (IC50 62.1 μM)
  • Shows cytotoxicity against african green monkey Vero cells (IC50 52.8 μM)

Catalog No. 50-004-18716 Supplier Medchemexpress LLC Supplier No. HYN39625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glycyrrhisoflavone, an active prenylflavonoid, inhibits α-glucosidase. It is intended for research use only.

  • Active prenylflavonoid
  • Inhibits α-glucosidase
  • For research use only
  • Purity of 99.43%
  • Appearance is off-white to light yellow solid
  • Classified as a flavonoid, isoflavone, phenol, polyphenol
  • Derived from Glycyrrhiza uralensis Fisch.
  • Shows cytotoxicity against human PC3 cells (IC50 62.1 μM)
  • Shows cytotoxicity against african green monkey Vero cells (IC50 52.8 μM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.