Learn More
Medchemexpress LLC Glycyrrhisoflavone | 116709-70-7 | 99.4% | 354.35 g·mol⁻¹ | C20H18O6 | 10 MG
SDP
Catalog No. 5000289847
Encompass_Preferred
Glycyrrhisoflavone is an active prenylflavonoid that inhibits α-glucosidase, provided as an analytical standard for research and analytical applications. It is supplied with defined purity and molecular characterization useful for assay development and analytical workflows.
  • High purity of 99.43%.
  • Provided as a 10 mg analytical standard.
  • Useful as an α-glucosidase inhibitor in biochemical assays.
  • Well-characterized molecular properties: C20H18O6, molecular weight 354.35 g·mol⁻¹.
  • CAS number 116709-70-7 for unambiguous identification.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-89847 Supplier Medchemexpress LLC Supplier No. HYN396210MG
Glycyrrhisoflavone is an active prenylflavonoid that inhibits α-glucosidase, provided as an analytical standard for research and analytical applications. It is supplied with defined purity and molecular characterization useful for assay development and analytical workflows.
  • High purity of 99.43%.
  • Provided as a 10 mg analytical standard.
  • Useful as an α-glucosidase inhibitor in biochemical assays.
  • Well-characterized molecular properties: C20H18O6, molecular weight 354.35 g·mol⁻¹.
  • CAS number 116709-70-7 for unambiguous identification.
  • For research use only; not for human or clinical use.