Learn More
Medchemexpress LLC Glycyrrhizic acid | 1405-86-3 | 99.8% | 822.93 | 50 MG
SDP
Catalog No. 5000366142
Encompass_Preferred
Glycyrrhizic acid is a triterpenoid saponin, functioning as a direct HMGB1 antagonist, exhibiting anti-tumor and anti-diabetic activities. It also offers several benefits for research applications.
  • Exhibits broad-spectrum anti-cancer activity.
  • Increases resistance to chemotherapy and radiation tissue toxicity.
  • Enhances drug absorption.
  • Possesses anti-multidrug resistance mechanisms.
  • Can act as a carrier material in drug delivery systems.
  • Promotes GLP-1 secretion in intestinal NCI-H716 cells through TGR5 activation.
  • Forms stable transparent low-molecular-weight hydrogels.
  • Increases plasma GLP-1 levels in type 1-like diabetic rats.
  • Significantly decreases levels of TgAb, HMGB1, TNF-α, IL-6, and IL-1β in mice.
Catalog No. 50-003-66142 Supplier Medchemexpress LLC Supplier No. 50MGHYN0184
Glycyrrhizic acid is a triterpenoid saponin, functioning as a direct HMGB1 antagonist, exhibiting anti-tumor and anti-diabetic activities. It also offers several benefits for research applications.
  • Exhibits broad-spectrum anti-cancer activity.
  • Increases resistance to chemotherapy and radiation tissue toxicity.
  • Enhances drug absorption.
  • Possesses anti-multidrug resistance mechanisms.
  • Can act as a carrier material in drug delivery systems.
  • Promotes GLP-1 secretion in intestinal NCI-H716 cells through TGR5 activation.
  • Forms stable transparent low-molecular-weight hydrogels.
  • Increases plasma GLP-1 levels in type 1-like diabetic rats.
  • Significantly decreases levels of TgAb, HMGB1, TNF-α, IL-6, and IL-1β in mice.