Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycyrrhizic acid ammonium salt, ≥90.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014537 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4537 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4537 Supplier MilliporeSigma Supelco Supplier No. 9144310MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard suitable for the quantitative determination of glycyrrhizic acid using high performance liquid chromatography.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53956-04-0
Molecular Weight (g/mol) 839.96
MDL Number MFCD00167400

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C42H62O16 · NH3
Quantity 10 mg
Formula Weight 839.96
Percent Purity ≥90.0% (HPLC)
Grade For HPLC
Physical Form Neat
ShelfLife Limited shelf life, expiry date on the label
RTECSNumber LZ6500000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.