Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glycyrrhizic Acid Ammonium Salt Fro mglycyrrhiza Root (Licorice)

Catalog No. G213725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G213725G 25 g
G2137100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G213725G Supplier Sigma Aldrich Supplier No. G213725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Glycyrrhizic acid (GA) is a non-selective inhibitor of 11+a-hydroxysteroid dehydrogenase (11+a-HSD), which mediates the inter-conversion of active and inactive glucocorticoids. GA also shows inhibitory effect on the cytokine activity of high-mobility group box 1 protein (HMGB1).The product has been used a source of glycyrrhizic acid (GA), to study its effect on glucose intolerance and insulin resistance in rats fed on high-fat/high-sucrose (HF/HS) diet.Biologically active constituent in the sweet root of Glycyrrhiza species (licorice). Antiviral.

Specifications

CAS 53956-04-0
Molecular Formula C42H65NO16
Linear Formula C42H62O16 -+ NH3
MDL Number MFCD00167400
Quantity 25 g
Synonym 3-O-(2-O-beta-D-Glucopyranuronosyl-alpha-D-glucopyranuronosyl)-18beta-glycyrrhetinic acid ammonium salt; Glycyrrhizin
Molecular Weight (g/mol) 839.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.