Learn More
Medchemexpress LLC Glycyrrhizic acid (standard) | 1405-86-3 | 97.5% | 10 MG
SDP
Catalog No. 5000295795
Encompass_Preferred
Glycyrrhizic acid (standard) is an analytical reference material of glycyrrhizic acid intended for method development, calibration, and compound identification in analytical workflows. Supplied as a solid in small mass packages, it is suitable for HPLC and LC-MS reference use. CAS 1405-86-3; molecular weight 822.93 g/mol; formula C42H62O16.
  • Intended for analytical applications such as HPLC and LC-MS calibration.
  • Provided as a solid reference standard in small mass packs (10 MG).
  • Identified by CAS 1405-86-3 and molecular weight 822.93 g/mol.
  • Used for quality control and compound identification.
  • Batch-specific purity and certificate of analysis available on request.
Catalog No. 50-002-95795 Supplier Medchemexpress LLC Supplier No. HYN0184R10MG
Glycyrrhizic acid (standard) is an analytical reference material of glycyrrhizic acid intended for method development, calibration, and compound identification in analytical workflows. Supplied as a solid in small mass packages, it is suitable for HPLC and LC-MS reference use. CAS 1405-86-3; molecular weight 822.93 g/mol; formula C42H62O16.
  • Intended for analytical applications such as HPLC and LC-MS calibration.
  • Provided as a solid reference standard in small mass packs (10 MG).
  • Identified by CAS 1405-86-3 and molecular weight 822.93 g/mol.
  • Used for quality control and compound identification.
  • Batch-specific purity and certificate of analysis available on request.