Learn More
Cayman Chemical Glycyrrhizic Acid (ammonium salt), Purity: ≥80%, CAS: 53956-04-0, 10 g
SDP
Catalog No. 502952285
Encompass_Preferred
A natural agonist of the human sweet taste receptor and used as a flavorant; has diverse cellular effects at low doses, including antioxidant, anti-inflammatory, antimicrobial, and antiviral actions
Catalog No. 50-295-2285 Supplier Cayman Chemical Supplier No. 1184710 G
A natural agonist of the human sweet taste receptor and used as a flavorant; has diverse cellular effects at low doses, including antioxidant, anti-inflammatory, antimicrobial, and antiviral actions