Learn More
Selleck Chemical LLC Glycyrrhizin (Glycyrrhizic Acid) 10mg 1405-86-3
SDP
Supplier:  Selleck Chemical LLC S230210MG
Encompass_Preferred
Glycyrrhizin (Glycyrrhizic Acid) is a direct HMGB1(high mobility group box 1) inhibitor that inhibits HMGB1-dependent inflammatory molecule expression and oxidative stress; modulates P38 and P-JNK but not p-ERK signalling; Also inhibits 11 beta-hydroxysteroid dehydrogenase and monoamine oxidase (MAO). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6165