Learn More
Selleck Chemical LLC Glycyrrhizin (NSC 167409) S2302-100mg
SDP
Catalog No. 503111787
Encompass_Preferred
Glycyrrhizin (NSC 167409 Glycyrrhizic Acid) is a direct HMGB1(high mobility group box 1) inhibitor that inhibits HMGB1-dependent inflammatory molecule expression and oxidative stress modulates P38 and P-JNK but not p-ERK signalling Also inhibits 11 beta-hydroxysteroid dehydrogenase and monoamine oxidase (MAO)
Catalog No. 50-311-1787 Supplier Selleck Chemical LLC Supplier No. S2302100MG
Glycyrrhizin (NSC 167409 Glycyrrhizic Acid) is a direct HMGB1(high mobility group box 1) inhibitor that inhibits HMGB1-dependent inflammatory molecule expression and oxidative stress modulates P38 and P-JNK but not p-ERK signalling Also inhibits 11 beta-hydroxysteroid dehydrogenase and monoamine oxidase (MAO)