Learn More
Apexbio Technology LLC GlyH-101 328541-79-3 10mg
SDP
Catalog No. 5000094138
Encompass_Preferred
GlyH-101 (CAS 328541-79-3) is a cell-permeable glycinyl hydrazone derivative that acts as a selective inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel exhibiting a Ki of 1 4 M By blocking CFTR-mediated chloride transport GlyH-101 serves as a valuable tool for investigating CFTR function in epithelial physiology and pathophysiology as well as for studying mechanisms underlying cystic fibrosis and related disorders involving chloride ion dysregulation
Catalog No. 50-000-94138 Supplier Apexbio Technology LLC Supplier No. B108710
GlyH-101 (CAS 328541-79-3) is a cell-permeable glycinyl hydrazone derivative that acts as a selective inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel exhibiting a Ki of 1 4 M By blocking CFTR-mediated chloride transport GlyH-101 serves as a valuable tool for investigating CFTR function in epithelial physiology and pathophysiology as well as for studying mechanisms underlying cystic fibrosis and related disorders involving chloride ion dysregulation