Learn More
Apexbio Technology LLC GlyH-101 328541-79-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089114
Encompass_Preferred
GlyH-101 (CAS 328541-79-3) is a cell-permeable glycinyl hydrazone derivative that acts as a selective inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel exhibiting a Ki of 1 4 M By blocking CFTR-mediated chloride transport GlyH-101 serves as a valuable tool for investigating CFTR function in epithelial physiology and pathophysiology as well as for studying mechanisms underlying cystic fibrosis and related disorders involving chloride ion dysregulation
Catalog No. 50-000-89114 Supplier Apexbio Technology LLC Supplier No. B10875.1
GlyH-101 (CAS 328541-79-3) is a cell-permeable glycinyl hydrazone derivative that acts as a selective inhibitor of the cystic fibrosis transmembrane conductance regulator (CFTR) chloride channel exhibiting a Ki of 1 4 M By blocking CFTR-mediated chloride transport GlyH-101 serves as a valuable tool for investigating CFTR function in epithelial physiology and pathophysiology as well as for studying mechanisms underlying cystic fibrosis and related disorders involving chloride ion dysregulation