Learn More
Medchemexpress LLC GlyH-101 | 328541-79-3 | 99.7% | 493.15 | 5 MG
SDP
Catalog No. 5000387598
Encompass_Preferred
GlyH-101 is a potent inhibitor of CFTR (Cystic Fibrosis Transmembrane conductance Regulator) and VSORC (Volume-Sensitive Outwardly Rectifying Chloride) conductance. It exhibits antiproliferative activity by inhibiting CFTR-like and VSORC currents and is intended for research use only.
  • Potent CFTR inhibitor
  • Potent and reversible VSORC inhibitor
  • Exhibits antiproliferative activity in PCT and PS120 cells
  • Inhibits CFTR-like current in PCT cells
  • Inhibits VSORC current in PS120 and PCT cells
  • Reduces cholera toxin-induced intestinal fluid secretion in mice
  • Available as a solid, light yellow to yellow
  • Soluble in DMSO
Catalog No. 50-003-87598 Supplier Medchemexpress LLC Supplier No. HY183365MG
GlyH-101 is a potent inhibitor of CFTR (Cystic Fibrosis Transmembrane conductance Regulator) and VSORC (Volume-Sensitive Outwardly Rectifying Chloride) conductance. It exhibits antiproliferative activity by inhibiting CFTR-like and VSORC currents and is intended for research use only.
  • Potent CFTR inhibitor
  • Potent and reversible VSORC inhibitor
  • Exhibits antiproliferative activity in PCT and PS120 cells
  • Inhibits CFTR-like current in PCT cells
  • Inhibits VSORC current in PS120 and PCT cells
  • Reduces cholera toxin-induced intestinal fluid secretion in mice
  • Available as a solid, light yellow to yellow
  • Soluble in DMSO