Learn More
Agilent Technologies AdvanceBio 2-AB alpha(2,3) Sialylated biantennary N-glycan library
Catalog No. NC1165871
Encompass
Library consists of biantennary oligosaccharides containing alpha(2,3) linked sialic acid. The reducing termini are derivatized with the fluorescent dye, 2-aminobenzamide (2-AB).
Catalog No. NC1165871 Supplier Agilent Technologies Supplier No. GKSB232
Library consists of biantennary oligosaccharides containing alpha(2,3) linked sialic acid. The reducing termini are derivatized with the fluorescent dye, 2-aminobenzamide (2-AB).