Learn More
Agilent Technologies GLYKO B (1-3 4) GALACTOSIDASE
Catalog No. NC0062648
Encompass
Hydrolyzes non-reducing terminal galactose β(1-3) and β(1-4) linkages. Can be used in conjunction with other β-galactosidases for exoglycosidase sequencing.
Catalog No. NC0062648 Supplier Agilent Technologies Supplier No. GKX5013
Hydrolyzes non-reducing terminal galactose β(1-3) and β(1-4) linkages. Can be used in conjunction with other β-galactosidases for exoglycosidase sequencing.