Learn More
Agilent Technologies AdvanceBio InstantPC G0F / FA2 N-Glycan Standard, 25 Injections
Catalog No. NC1286954
Encompass
The AdvanceBio InstantPC G0F / FA2 N-Glycan Standard is a high-purity, complex-type native N-glycan standard designed for analytical chemical laboratory use. It serves as a reliable reference material for glycan analysis applications.
  • Supplied as a whitish powder
  • Soluble in water
  • Monoisotopic mass of 1723.6922
  • Suitable for complex-type native N-glycan identification
  • Formulated for analytical applications
  • Quantity provides 25 injections
Catalog No. NC1286954 Supplier Agilent Technologies Supplier No. GKPC302
The AdvanceBio InstantPC G0F / FA2 N-Glycan Standard is a high-purity, complex-type native N-glycan standard designed for analytical chemical laboratory use. It serves as a reliable reference material for glycan analysis applications.
  • Supplied as a whitish powder
  • Soluble in water
  • Monoisotopic mass of 1723.6922
  • Suitable for complex-type native N-glycan identification
  • Formulated for analytical applications
  • Quantity provides 25 injections